missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-fluorophenylacetate, 99%, Thermo Scientific Chemicals

Product Code. 11335649
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11335649 5 g 5g
11345649 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11335649 Supplier Thermo Scientific Alfa Aesar Supplier No. L19999.06
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 584-74-7 | Molecular Formula: C10H11FO2 | Molecular Weight: 182.194 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 584-74-7
Molecular Formula C10H11FO2
Molecular Weight (g/mol) 182.194
MDL Number MFCD04039343
InChI Key OFWMNCYZASDVSS-UHFFFAOYSA-N
Synonym ethyl 2-2-fluorophenyl acetate, ethyl 2-fluorophenylacetate, ethyl 2-fluorophenyl acetate, 2-fluorophenylacetic acid ethyl ester, benzeneacetic acid, 2-fluoro-, ethyl ester, acmc-209uz7, ethyl 2-fluorobenzeneacetate
PubChem CID 7578179
IUPAC Name ethyl 2-(2-fluorophenyl)acetate
SMILES CCOC(=O)CC1=CC=CC=C1F

Specifications

Density 1.101
Boiling Point 235°C
Flash Point 85°C (185°F)
Refractive Index 1.482
Quantity 5 g
Formula Weight 182.2
Percent Purity 99%
Chemical Name or Material Ethyl 2-fluorophenylacetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.