missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-fluoroisobutyrate, 97%, Thermo Scientific Chemicals

Product Code. 15455717
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15455717 5 g 5g
15445717 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15455717 Supplier Thermo Scientific Alfa Aesar Supplier No. H32931.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 55816-69-8 | Molecular Formula: C6H11FO2 | Molecular Weight: 134.15 g/mol

Ethyl 2-fluoroisobutyrate is used as an active pharmaceutical ingredient.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 2-fluoroisobutyrate is used as an active pharmaceutical ingredient.

Solubility
Not miscible or difficult to mix with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55816-69-8
Molecular Formula C6H11FO2
Molecular Weight (g/mol) 134.15
MDL Number MFCD08062432
InChI Key CJRQQJKWNULSFQ-UHFFFAOYSA-N
Synonym ethyl 2-fluoroisobutyrate, propanoic acid, 2-fluoro-2-methyl-, ethyl ester, ethyl 2-fluoro-isobutyrate, ethyl 2-fluoro-2-methylpropionate, 2-fluoro-2-methylpropionic acid ethyl ester
PubChem CID 10898763
IUPAC Name ethyl 2-fluoro-2-methylpropanoate
SMILES CCOC(=O)C(C)(C)F

Specifications

Boiling Point 122°C to 126°C
Quantity 5 g
UN Number UN1992
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 134.15
Percent Purity 97%
Chemical Name or Material Ethyl 2-fluoroisobutyrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.