missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-ethylacetoacetate, 95%

Product Code. 15493237
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15493237 25 g 25g
15403247 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15493237 Supplier Thermo Scientific Alfa Aesar Supplier No. B22295.14
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 607-97-6 | C8H14O3 | 158.197 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 607-97-6
Molecular Formula C8H14O3
Molecular Weight (g/mol) 158.197
MDL Number MFCD00039898
InChI Key OKANYBNORCUPKZ-UHFFFAOYSA-N
Synonym ethyl 2-ethylacetoacetate, ethyl 2-acetylbutyrate, butanoic acid, 2-ethyl-3-oxo-, ethyl ester, 2-ethylacetoacetic acid ethyl ester, ethyl 2-ethyl-3-ketobutyrate, ethyl alpha-ethylacetoacetate, ethyl-2-ethylacetoacetate, 2-ethyl-3-oxo-butyric acid ethyl ester, ethyl .alpha.-acetylbutyrate, ethyl 2-ethyl-3-oxo-butanoate
PubChem CID 79065
IUPAC Name ethyl 2-ethyl-3-oxobutanoate
SMILES CCC(C(=O)C)C(=O)OCC

Specifications

Density 1.009
Boiling Point 83°C to 85°C (10 mmHg)
Flash Point 77°C (170°F)
Refractive Index 1.422
Quantity 25 g
Beilstein 636286
Formula Weight 158.2
Percent Purity 95%
Chemical Name or Material Ethyl 2-ethylacetoacetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.