missing translation for 'onlineSavingsMsg'
Learn More

Ethyl [2-Diethylaminothiocarboxyl)]phenylacetate, TRC

Product Code. 30373247
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30373247 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30373247 Supplier Toronto Research Chemicals Supplier No. TRCE916450100MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl [2-Diethylaminothiocarboxyl]phenylacetate
CAS 1076198-04-3
Molecular Formula C15H21NO3S
Quantity 100 mg
Synonym 2-[(Diethylamino)thioxomethoxy]benzeneacetic Acid Ethyl Ester
InChI Formula InChI=1S/C15H21NO3S/c1-4-16(5-2)15(20)19-13-10-8-7-9-12(13)11-14(17)18-6-3/h7-10H,4-6,11H2,1-3H3
SMILES CCOC(=O)Cc1ccccc1OC(=S)N(CC)CC
IUPAC Name ethyl 2-[2-(diethylcarbamothioyloxy)phenyl]acetate
Molecular Weight (g/mol) 295.4
Formula Weight 295.1242
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.