missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-cyanoacrylate

Product Code. 11975621 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 11975621

Brand: Thermo Scientific Acros 434450050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7085-85-0 | C6H7NO2 | 125.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7085-85-0
Melting Point -20.0°C
Color Colorless to Yellow
Density 1.0400g/mL
Boiling Point 54.0°C to 56.0°C
Flash Point 85°C
Packaging Glass bottle
Molecular Formula C6H7NO2
Linear Formula CNC(=CH2)COOCH2CH3
MDL Number MFCD00045615
Quantity 5 g
Merck Index 15,3842
Specific Gravity 1.04
Synonym ethyl 2-cyanoacrylate, ethyl cyanoacrylate, 2-propenoic acid, 2-cyano-, ethyl ester, unii-2g95foh7sf, ccris 1693, acrylic acid, 2-cyano-, ethyl ester, 2g95foh7sf, 2-cyanoacrylic acid ethyl ester, 2-propenoic acid,2-cyano-, ethyl ester, dsstox_cid_5279
Solubility Information Solubility in water: reacts. Other solubilities: soluble in methyl ethyl ketone, toluene, acetone,, dmf, nitromethane
InChI Key FGBJXOREULPLGL-UHFFFAOYSA-N
SMILES CCOC(=O)C(=C)C#N
IUPAC Name ethyl 2-cyanoprop-2-enoate
Molecular Weight (g/mol) 125.13
PubChem CID 81530
Formula Weight 125.13
Concentration or Composition (by Analyte or Components) may contain varying amounts of polymerized material
Physical Form Liquid
Chemical Name or Material Ethyl 2-cyanoacrylate
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl 2-cyanoacrylate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.