missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-cyanoacetoacetate

Product Code. 11380428
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11380428 2 g 2g
11390428 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11380428 Supplier Thermo Scientific Alfa Aesar Supplier No. 043792.04
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 634-55-9 | C7H9NO3 | 155.15 g/mol

It is used as a pharmaceutical intermediate. It is also used in the synthesis of fluorine-containing alkyl 2-cyano-3-oxocarboxylates and also in the syntheses of pyrazolo [1, 2-a] pyrazole and pyrazolo [5, 1-b] [1, 3] oxazine derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 634-55-9
Molecular Formula C7H9NO3
Molecular Weight (g/mol) 155.15
MDL Number MFCD00043487
InChI Key NWOKVFOTWMZMHL-UHFFFAOYNA-N
Synonym ethyl 2-cyanoacetoacetate, ethyl 2-acetylcyanoacetate, ethyl a-cyanoacetoacetate, ethyl alpha-cyanoacetoacetate, butanoic acid, 2-cyano-3-oxo-, ethyl ester, ethyl2-acetylcyanoacetate, ethyl 2-cyano-3-oxo-butanoate, 2-cyanoacetoacetic acid ethyl ester, 2-cyano-3-oxo-butyric acid ethyl ester, zlchem 939
PubChem CID 136455
ChEBI CHEBI:51924
IUPAC Name ethyl 2-cyano-3-oxobutanoate
SMILES CCOC(=O)C(C#N)C(C)=O

Specifications

Quantity 2 g
UN Number UN3276
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 155.16
Physical Form Liquid
Chemical Name or Material Ethyl 2-cyanoacetoacetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.