missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-Chloropropionate, TRC

Product Code. 30380775
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380775 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30380775 Supplier Toronto Research Chemicals Supplier No. TRCE90362010G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Chloropropionic acid-ethyl ester
CAS 535-13-7
Molecular Formula C5 H9 Cl O2
Quantity 10 g
Synonym Propionic acid, 2-chloro-, ethyl ester (6CI,7CI,8CI), (2RS)-2-Chloropropionic acid ethyl ester, 2-Chloropropanoic acid ethyl ester, Ethyl 2-chloropropanoate, Ethyl 2-chloropropionate, Ethyl α-chloropropionate, NSC 522671
InChI Formula InChI=1S/C5H9ClO2/c1-3-8-5(7)4(2)6/h4H,3H2,1-2H3
SMILES CCOC(=O)C(C)Cl
IUPAC Name ethyl 2-chloropropanoate
Molecular Weight (g/mol) 136.58
Formula Weight 136.0291
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.