missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-chloropropionate, 97%, Thermo Scientific™

Product Code. 11383037
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11383037 250 g 250g
11373037 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11383037 Supplier Thermo Scientific Alfa Aesar Supplier No. L13410.30

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 535-13-7
Molecular Formula C5H9ClO2
Molecular Weight (g/mol) 136.58
MDL Number MFCD00000864
InChI Key JEAVBVKAYUCPAQ-UHFFFAOYNA-N
Synonym ethyl 2-chloropropionate, 2-chloropropionic acid ethyl ester, propanoic acid, 2-chloro-, ethyl ester, ethyl-2-chloropropionate, 2-chloropropionic acid, ethyl ester, ethyl alpha-chloropropionate, ethyl .alpha.-chloropropionate, propionic acid, 2-chloro-, ethyl ester, +-ethyl 2-chloropropionate, ethyl ?-chloropropionate
PubChem CID 10807
IUPAC Name ethyl 2-chloropropanoate
SMILES CCOC(=O)C(C)Cl

Specifications

Density 1.072
Boiling Point 146°C to 149°C
Flash Point 38°C (100°F)
Odor Pungent
Refractive Index 1.417
Quantity 250 g
UN Number UN2935
Beilstein 773906
Merck Index 14,3785
Formula Weight 136.58
Percent Purity 97%
Chemical Name or Material Ethyl 2-chloropropionate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.