missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-chlorooxazole-4-carboxylate, 95%, Thermo Scientific Chemicals

Product Code. p-7030695
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15474117 250 mg 250mg
11355135 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15474117 Supplier Thermo Scientific Alfa Aesar Supplier No. H31521.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 460081-18-9
Molecular Formula C6H6ClNO3
Molecular Weight (g/mol) 175.568
MDL Number MFCD06660120
InChI Key SYWQOPRAPDMWMC-UHFFFAOYSA-N
PubChem CID 2763184
IUPAC Name ethyl 2-chloro-1,3-oxazole-4-carboxylate
SMILES CCOC(=O)C1=COC(=N1)Cl

Specifications

Melting Point 97°C to 98°C
Quantity 250 mg
Synonym ethyl 2-chlorooxazole-4-carboxylate, 2-chlorooxazole-4-carboxylic acid ethyl ester, ethyl2-chlorooxazole-4-carboxylate, ethyl 2-chloroxazole-4-carboxylate, ethyl-2-chlorooxazole-4-carboxylate, 2-chloro-oxazole-4-carboxylic acid ethyl ester, 4-oxazolecarboxylic acid, 2-chloro-, ethyl ester, 2-chloro-4-oxazolecarboxylic acid ethyl ester, 2-chloro-1,3-ixozole-4-carboxylic acid ethyl ester, hodgetts reagent
Formula Weight 175.57
Percent Purity 95%
Chemical Name or Material Ethyl 2-chlorooxazole-4-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.