missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-chlorobenzoate, 98%

Product Code. 11409412
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11409412 100 g 100g
11499402 25 g 25g
11419412 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11409412 Supplier Thermo Scientific Alfa Aesar Supplier No. A10141.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7335-25-3 | C9H9ClO2 | 184.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7335-25-3
Molecular Formula C9H9ClO2
Molecular Weight (g/mol) 184.62
MDL Number MFCD00016338
InChI Key RETLCWPMLJPOTP-UHFFFAOYSA-N
Synonym 2-chlorobenzoic acid ethyl ester, ethyl o-chlorobenzoate, benzoic acid, 2-chloro-, ethyl ester, ethyl-2-chlorobenzoate, ethyl2-chlorobenzoate, pubchem3703, acmc-1bfxj, ethyl 2-chloranylbenzoate, chlorobenzoic acid ethyl ester, ksc498a5r
PubChem CID 81783
IUPAC Name ethyl 2-chlorobenzoate
SMILES CCOC(=O)C1=CC=CC=C1Cl

Specifications

Density 1.194
Boiling Point 243°C
Flash Point >110°C (230°F)
Refractive Index 1.524
Quantity 100 g
Beilstein 2207359
Formula Weight 184.62
Percent Purity 98%
Chemical Name or Material Ethyl 2-chlorobenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.