missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-chloroacetoacetate, 90%

Product Code. 10520413 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10520413 250 mL Glass bottle 250mL
10361213 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10520413 Supplier Thermo Scientific Acros Supplier No. 376312500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 609-15-4 | C6H9ClO3 | 164.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 609-15-4
Molecular Formula C6H9ClO3
Molecular Weight (g/mol) 164.59
MDL Number MFCD00009141
InChI Key RDULEYWUGKOCMR-UHFFFAOYNA-N
PubChem CID 11858
IUPAC Name ethyl 2-chloro-3-oxobutanoate
SMILES CCOC(=O)C(Cl)C(C)=O

Specifications

Melting Point -80°C
Color Yellow
Density 1.1900g/mL
Boiling Point 200°C
Flash Point 54°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Linear Formula CH3COCH(Cl)CO2C2H5
Refractive Index 1.4410 to 1.444
Quantity 250 mL
Specific Gravity 1.19
Synonym ethyl 2-chloroacetoacetate, 2-chloroacetoacetic acid ethyl ester, butanoic acid, 2-chloro-3-oxo-, ethyl ester, ethyl alpha-chloroacetoacetate, ethyl-2-chloracetoacetate, 2-chloro-3-oxobutyric acid ethyl ester, ethyl2-chloroacetoacetate, 2-chloro-3-oxobutanoic acid, ethyl ester, ethyl2-chloro-3-oxobutanoate, ethyl 2-chloro-3-oxobutyrate
Solubility Information Solubility in water: 17g/L (20°C)
Formula Weight 164.59
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material Ethyl 2-chloroacetoacetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.