missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-chloro-4-methyl-1,3-thiazole-5-carboxylate, 97%, Thermo Scientific™

Product Code. 10127302
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10127302 250 mg 250mg
10516693 1 g 1g
10492674 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10127302 Supplier Thermo Scientific Maybridge Supplier No. MO07617CB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 7238-62-2
Molecular Formula C7H8ClNO2S
Molecular Weight (g/mol) 205.656
InChI Key VUARUZUFHDNJSY-UHFFFAOYSA-N
Synonym ethyl 2-chloro-4-methylthiazole-5-carboxylate, ethyl 2-chloro-4-methyl-5-thiazolecarboxylate, 5-thiazolecarboxylic acid, 2-chloro-4-methyl-, ethyl ester, 2-chloro-4-methyl-5-thiazolecarboxylic acid ethyl ester, 5-thiazolecarboxylicacid, 2-chloro-4-methyl-, ethyl ester, 2-chloro-4-methyl-thiazole-5-carboxylic acid ethyl ester
PubChem CID 594903
IUPAC Name ethyl 2-chloro-4-methyl-1,3-thiazole-5-carboxylate
SMILES CCOC(=O)C1=C(N=C(S1)Cl)C

Specifications

Assay Percent Range 97%
Quantity 250 mg
Formula Weight 205.66
Percent Purity 97%
Chemical Name or Material Ethyl 2-chloro-4-methyl-1, 3-thiazole-5-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.