missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-chloro-4,4,4-trifluoroacetoacetate, 94%

Product Code. 11485926
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11485926 25 g 25g
11475926 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11485926 Supplier Thermo Scientific Alfa Aesar Supplier No. B22270.14
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 363-58-6 | C6H6ClF3O3 | 218.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 363-58-6
Molecular Formula C6H6ClF3O3
Molecular Weight (g/mol) 218.56
MDL Number MFCD00041540
InChI Key YVWUNJVPOCYLIM-VKHMYHEASA-N
Synonym ethyl 2-chloro-4,4,4-trifluoroacetoacetate, ethyl 2-chloro-3-keto-4,4,4-trifluorobutyrate, ethyl 2-chloro-4,4,4-trifluoro-3-oxobutyrate, 2-chloro-4,4,4-trifluoro-3-oxo-butyric acid ethyl ester, 2-chloro-4,4,4-trifluoroacetoacetic acid ethyl ester, butanoic acid, 2-chloro-4,4,4-trifluoro-3-oxo-, ethyl ester, zlchem 1230, acmc-1cs5z, ethyl chlorotrifluoroacetoacetate, ethyl 2-chloro-3-keto-trifluorobutyrate
PubChem CID 2737165
IUPAC Name ethyl 2-chloro-4,4,4-trifluoro-3-oxobutanoate
SMILES CCOC(=O)[C@@H](Cl)C(=O)C(F)(F)F

Specifications

Density 1.39
Boiling Point 67°C to 71°C (35 mmHg)
Flash Point 67°C (153°F)
Refractive Index 1.39
Quantity 25 g
UN Number UN1760
Beilstein 1787023
Formula Weight 218.56
Percent Purity 94%
Chemical Name or Material Ethyl 2-chloro-4,4,4-trifluoroacetoacetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.