missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-chloro-4,4,4-trifluoroacetoacetate, 94%

Product Code. 11475926
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11475926

Brand: Thermo Scientific Alfa Aesar B22270.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 363-58-6 | C6H6ClF3O3 | 218.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 363-58-6
Molecular Formula C6H6ClF3O3
Molecular Weight (g/mol) 218.56
MDL Number MFCD00041540
InChI Key YVWUNJVPOCYLIM-VKHMYHEASA-N
Synonym ethyl 2-chloro-4,4,4-trifluoroacetoacetate, ethyl 2-chloro-3-keto-4,4,4-trifluorobutyrate, ethyl 2-chloro-4,4,4-trifluoro-3-oxobutyrate, 2-chloro-4,4,4-trifluoro-3-oxo-butyric acid ethyl ester, 2-chloro-4,4,4-trifluoroacetoacetic acid ethyl ester, butanoic acid, 2-chloro-4,4,4-trifluoro-3-oxo-, ethyl ester, zlchem 1230, acmc-1cs5z, ethyl chlorotrifluoroacetoacetate, ethyl 2-chloro-3-keto-trifluorobutyrate
PubChem CID 2737165
IUPAC Name ethyl 2-chloro-4,4,4-trifluoro-3-oxobutanoate
SMILES CCOC(=O)[C@@H](Cl)C(=O)C(F)(F)F

Specifications

Density 1.39
Boiling Point 67°C to 71°C (35 mmHg)
Flash Point 67°C (153°F)
Refractive Index 1.39
Quantity 5 g
UN Number UN1760
Beilstein 1787023
Formula Weight 218.56
Percent Purity 94%
Chemical Name or Material Ethyl 2-chloro-4,4,4-trifluoroacetoacetate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.