missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-chloro-3-pyridylglyoxylate, 97%, Thermo Scientific Chemicals

Catalog Number 11459270
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
902837-56-3
C9H8ClNO3
213.617
MFCD07781194
ZTIMVJIGNBYGJG-UHFFFAOYSA-N
ethyl 2-chloro-3-pyridylglyoxylate, ethyl 2-2-chloropyridin-3-yl-2-oxoacetate, ethyl 2-chloropyridin-3-yl oxo acetate, ethyl 2-chloro-3-pyridinyl oxoacetate, 2-chloro-3-pyridylglyoxylic acid ethyl ester, ethyl 2-2-chloro 3-pyridyl-2-oxoacetate, 2-chloro-pyridin-3-yl-oxo-acetic acid ethyl ester
42553166
ethyl 2-(2-chloropyridin-3-yl)-2-oxoacetate
CCOC(=O)C(=O)C1=C(N=CC=C1)Cl
This item is not returnable. View return policy
902837-56-3
C9H8ClNO3
213.617
MFCD07781194
ZTIMVJIGNBYGJG-UHFFFAOYSA-N
ethyl 2-chloro-3-pyridylglyoxylate, ethyl 2-2-chloropyridin-3-yl-2-oxoacetate, ethyl 2-chloropyridin-3-yl oxo acetate, ethyl 2-chloro-3-pyridinyl oxoacetate, 2-chloro-3-pyridylglyoxylic acid ethyl ester, ethyl 2-2-chloro 3-pyridyl-2-oxoacetate, 2-chloro-pyridin-3-yl-oxo-acetic acid ethyl ester
42553166
ethyl 2-(2-chloropyridin-3-yl)-2-oxoacetate
CCOC(=O)C(=O)C1=C(N=CC=C1)Cl

Ethyl 2-chloro-3-pyridylglyoxylate is used as a pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 2-chloro-3-pyridylglyoxylate is used as a pharmaceutical intermediates.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 213.62
Percent Purity 97%
Chemical Name or Material Ethyl 2-chloro-3-pyridylglyoxylate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'