missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-Chloro-2-(2-(4-methoxyphenyl)hydrazono)acetate, TRC

Product Code. 30456444
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30456444 500 mg 500mg
30449620 100 mg 100mg
30453565 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30456444 Supplier Toronto Research Chemicals Supplier No. TRCZ346018500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27143-07-3
Molecular Formula C11H13N2O3Cl
Molecular Weight (g/mol) 256.68
IUPAC Name ethyl 2-chloro-2-[(4-methoxyphenyl)hydrazinylidene]acetate
SMILES CCOC(=O)C(=NNc1ccc(OC)cc1)Cl

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C11H13ClN2O3/c1-3-17-11(15)10(12)14-13-8-4-6-9(16-2)7-5-8/h4-7,13H,3H2,1-2H3
Formula Weight 256.0615
Chemical Name or Material Ethyl 2-Chloro-2-(2-(4-methoxyphenyl)hydrazono)acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.