missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-chloro-2-(2-(4-chlorophenyl)hydrazono)acetate, TRC

Product Code. 30384786
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30384786 100 mg
30372006 10 mg
30368813 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30384786 Supplier Toronto Research Chemicals Supplier No. TRCE903738100MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27143-09-5
Molecular Formula C10H10Cl2N2O2
Molecular Weight (g/mol) 261.1
Synonym 2-Chloro-2-[[(4-chlorophenyl)amino]imino]acetic Acid Ethyl Ester, Ethyl (4-chlorophenylhydrazono)chloromethylcarboxylate, Ethyl 2-chloro-2-[N'-(4-chlorophenyl)hydrazono]acetate
SMILES CCOC(=O)\C(=N\Nc1ccc(Cl)cc1)\Cl

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C10H10Cl2N2O2/c1-2-16-10(15)9(12)14-13-8-5-3-7(11)4-6-8/h3-6,13H,2H2,1H3/b14-9-
Formula Weight 260.0119
Chemical Name or Material Ethyl 2-chloro-2-(2-(4-chlorophenyl)hydrazono)acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.