missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-butynoate, 98%

Product Code. 11329899
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11329899 1 g 1g
11339899 5 g 5g
11349899 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11329899 Supplier Thermo Scientific Alfa Aesar Supplier No. B25370.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4341-76-8 | C6H8O2 | 112.128 g/mol

Ethyl 2-butynoate is used as a precursor for the synthesis of 3-ethyl, 1-methyl 1-(2-nitrophenyl)-cyclopent-3-ene-1,3-dicarboxylate, alkenylsilanols and tricyclic aziridine derivatives. It acts as a methanogenesis inhibitor. As an electron deficient alkyne derivative, it plays a vital role in the preparation of 1,3-dienes catalyzed by rhodium(I)/H8-BINAP complex.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 2-butynoate is used as a precursor for the synthesis of 3-ethyl, 1-methyl 1-(2-nitrophenyl)-cyclopent-3-ene-1,3-dicarboxylate, alkenylsilanols and tricyclic aziridine derivatives. It acts as a methanogenesis inhibitor. As an electron deficient alkyne derivative, it plays a vital role in the preparation of 1,3-dienes catalyzed by rhodium(I)/H8-BINAP complex.

Solubility
Miscible with water, chloroform and ethyl acetate.

Notes
Air sensitive. Incompatible with oxidizing agents, bases and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4341-76-8
Molecular Formula C6H8O2
Molecular Weight (g/mol) 112.128
MDL Number MFCD00015182
InChI Key FCJJZKCJURDYNF-UHFFFAOYSA-N
Synonym ethyl 2-butynoate, ethyl tetrolate, 2-butynoic acid ethyl ester, but-2-ynoic acid ethyl ester, 2-butynoic acid, ethyl ester, tetrolic acid ethyl ester, ethyl-2-butynoate, ethyl 3-methylpropiolate, acmc-1aeo2, ethyl 1-propynecarboxylate
PubChem CID 78043
IUPAC Name ethyl but-2-ynoate
SMILES CCOC(=O)C#CC

Specifications

Density 0.967
Boiling Point 163°C
Flash Point 58°C (127°F)
Odor Characteristic
Linear Formula CH3C≡CCO2CH2CH3
Refractive Index 1.438
Quantity 1 g
UN Number UN3272
Beilstein 1744948
Sensitivity Air Sensitive
Solubility Information Miscible with water,chloroform and ethyl acetate.
Formula Weight 112.13
Percent Purity 98%
Chemical Name or Material Ethyl 2-butynoate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.