missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-bromovalerate, 97%

Product Code. 11497206
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11497206 25 g 25g
11407216 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11497206 Supplier Thermo Scientific Alfa Aesar Supplier No. B25530.14
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 615-83-8 | C7H13BrO2 | 209.083 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 615-83-8
Molecular Formula C7H13BrO2
Molecular Weight (g/mol) 209.083
MDL Number MFCD00000159
InChI Key ORSIRXYHFPHWTN-UHFFFAOYSA-N
PubChem CID 79071
IUPAC Name ethyl 2-bromopentanoate
SMILES CCCC(C(=O)OCC)Br

Specifications

Density 1.226
Boiling Point 190°C to 192°C
Flash Point 77°C (170°F)
Refractive Index 1.448
Quantity 25 g
UN Number UN3265
Beilstein 1098901
Synonym ethyl 2-bromovalerate, ethyl2-bromovalerate, ethyl .alpha.-bromovalerate, pentanoic acid, 2-bromo-, ethyl ester, 2-bromovaleric acid ethyl ester, 2-bromopentanoic acid ethyl ester, 2-bromo-pentanoic acid ethyl ester, pentanoic acid, bromo-, ethyl ester, ethyl bromovalerate, ethyl-2bromovalerate
Formula Weight 209.09
Percent Purity 97%
Chemical Name or Material Ethyl 2-bromovalerate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.