missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-bromothiazole-4-carboxylate, 97%, Thermo Scientific™

Product Code. 15424107
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 15424107

Brand: Thermo Scientific Alfa Aesar H31507.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 100367-77-9
Melting Point 68°C to 71°C
Molecular Formula C6H6BrNO2S
MDL Number MFCD03788564
Quantity 1 g
Sensitivity Light and moisture sensitive
Synonym ethyl 2-bromothiazole-4-carboxylate, 2-bromothiazole-4-carboxylic acid ethyl ester, 4-thiazolecarboxylic acid, 2-bromo-, ethyl ester, ethyl2-bromothiazole-4-carboxylate, ethyl 2-bromo-4-thiazolecarboxylate, 2-bromo-thiazole-4-carboxylic acid ethyl ester, 2-bromo-4-ethoxycarbonyl-1,3-thiazole, 2-bromo-1,3-thiazole-4-carboxylic acid ethyl ester, zlchem 490, pubchem17466
InChI Key CNHISCQPKKGDPO-UHFFFAOYSA-N
SMILES CCOC(=O)C1=CSC(=N1)Br
IUPAC Name ethyl 2-bromo-1,3-thiazole-4-carboxylate
Molecular Weight (g/mol) 236.083
PubChem CID 353965
Formula Weight 236.08
Percent Purity 97%
Chemical Name or Material Ethyl 2-bromothiazole-4-carboxylate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.