missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-bromothiazole-4-carboxylate, 96%

Product Code. 15214378 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15214378 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15214378 Supplier Thermo Scientific Acros Supplier No. 454610010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 100367-77-9 | C6H6BrNO2S | 236.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 100367-77-9
Melting Point 69.0°C to 71.0°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass Bottle
Molecular Formula C6H6BrNO2S
Quantity 1 g
Synonym ethyl 2-bromothiazole-4-carboxylate, 2-bromothiazole-4-carboxylic acid ethyl ester, 4-thiazolecarboxylic acid, 2-bromo-, ethyl ester, ethyl2-bromothiazole-4-carboxylate, ethyl 2-bromo-4-thiazolecarboxylate, 2-bromo-thiazole-4-carboxylic acid ethyl ester, 2-bromo-4-ethoxycarbonyl-1,3-thiazole, 2-bromo-1,3-thiazole-4-carboxylic acid ethyl ester, zlchem 490, pubchem17466
InChI Key CNHISCQPKKGDPO-UHFFFAOYSA-N
SMILES CCOC(=O)C1=CSC(=N1)Br
IUPAC Name ethyl 2-bromo-1,3-thiazole-4-carboxylate
Molecular Weight (g/mol) 236.09
PubChem CID 353965
Formula Weight 236.09
Percent Purity 96%
Chemical Name or Material Ethyl 2-bromothiazole-4-carboxylate
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl 2-bromothiazole-4-carboxylate, 96%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.