missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-Bromopropionate, TRC

Product Code. 30380981
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380981 100 g -
30383509 25 g -
30384035 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30380981 Supplier Toronto Research Chemicals Supplier No. TRCE900470100G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 535-11-5
Molecular Formula C5H9BrO2
Molecular Weight (g/mol) 181.03
Synonym Ethyl DL-2-bromopropionate, Ethyl dl-alpha-bromopropionate, Ethyl alpha-bromopropanoate, Ethyl alpha-bromopropionate, NSC 6753, alpha-Bromopropionic Acid Ethyl Ester, (±)-2-Bromopropanoic Acid Ethyl Ester, (±)-2-Bromopropionic Acid Ethyl Ester, (±)-Ethyl alpha-bromopropionate, 2-Bromopropanoic Acid Ethyl Ester, 2-Bromopro pionic Acid Ethyl Ester, Ethyl (±)-2-bromopropionate, Ethyl 2-bromopropanoate, alpha-bromopropionic Acid Ethyl Ester, 2-Bromopropionic Acid Ethyl Ester
IUPAC Name ethyl 2-bromopropanoate
SMILES CCOC(=O)C(C)Br

Specifications

Quantity 100 g
InChI Formula InChI=1S/C5H9BrO2/c1-3-8-5(7)4(2)6/h4H,3H2,1-2H3
Formula Weight 179.9786
Chemical Name or Material Ethyl 2-Bromopropionate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.