missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-bromopropionate, 98+%, Thermo Scientific Chemicals

Product Code. 11448950
Click to view available options
Quantity:
50 g
250 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It’s an important organic intermediate can be used in agrochemical, pharmaceutical synthesis and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It’s an important organic intermediate can be used in agrochemical, pharmaceutical synthesis and dyestuff field.

Solubility
Immiscible in water. Soluble in alcohol and ether.

Notes
Store in dark and away from oxidizing agents and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 535-11-5
Molecular Formula C5H9BrO2
Molecular Weight (g/mol) 181.03
MDL Number MFCD00000144
InChI Key ARFLASKVLJTEJD-UHFFFAOYNA-N
Synonym ethyl 2-bromopropionate, ethyl alpha-bromopropionate, ethyl-2-bromopropionate, 2-bromopropanoic acid ethyl ester, propanoic acid, 2-bromo-, ethyl ester, alpha-bromopropionic acid ethyl ester, ethyl a-bromopropionate, dl-ethyl 2-bromopropionate, 2-bromopropionic acid ethyl ester, ethyl alpha-bromopropanoate
PubChem CID 79040
IUPAC Name ethyl 2-bromopropanoate
SMILES CCOC(=O)C(C)Br

Specifications

Density 1.391
Boiling Point 158°C to 160°C
Flash Point 51°C (123°F)
Refractive Index 1.446
Quantity 50 g
UN Number UN1992
Beilstein 773920
Merck Index 14,3772
Solubility Information Immiscible in water. Soluble in alcohol and ether.
Formula Weight 181.03
Percent Purity ≥98%
Chemical Name or Material Ethyl 2-bromopropionate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'