missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-bromopropionate, 98+%

Product Code. 11458950
Click to view available options
Quantity:
50 g
250 g
500 g
Unit Size:
250g
500g
50g
This item is not returnable. View return policy

Product Code. 11458950

Brand: Thermo Scientific Alfa Aesar A10820.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 535-11-5 | C5H9BrO2 | 181.03 g/mol

It’s an important organic intermediate can be used in agrochemical, pharmaceutical synthesis and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It’s an important organic intermediate can be used in agrochemical, pharmaceutical synthesis and dyestuff field.

Solubility
Immiscible in water. Soluble in alcohol and ether.

Notes
Store in dark and away from oxidizing agents and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 535-11-5
Molecular Formula C5H9BrO2
Molecular Weight (g/mol) 181.03
MDL Number MFCD00000144
InChI Key ARFLASKVLJTEJD-UHFFFAOYNA-N
Synonym ethyl 2-bromopropionate, ethyl alpha-bromopropionate, ethyl-2-bromopropionate, 2-bromopropanoic acid ethyl ester, propanoic acid, 2-bromo-, ethyl ester, alpha-bromopropionic acid ethyl ester, ethyl a-bromopropionate, dl-ethyl 2-bromopropionate, 2-bromopropionic acid ethyl ester, ethyl alpha-bromopropanoate
PubChem CID 79040
IUPAC Name ethyl 2-bromopropanoate
SMILES CCOC(=O)C(C)Br

Specifications

Density 1.391
Boiling Point 158°C to 160°C
Flash Point 51°C (123°F)
Refractive Index 1.446
Quantity 250 g
UN Number UN1992
Beilstein 773920
Merck Index 14,3772
Solubility Information Immiscible in water. Soluble in alcohol and ether.
Formula Weight 181.03
Percent Purity ≥98%
Chemical Name or Material Ethyl 2-bromopropionate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.