missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-bromonicotinate, 96%

Product Code. 15438987
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15438987

Brand: Thermo Scientific Alfa Aesar H52335.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 53087-78-8 | C8H8BrNO2 | 230.061 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53087-78-8
Molecular Formula C8H8BrNO2
Molecular Weight (g/mol) 230.061
MDL Number MFCD04113825
InChI Key QOFKYVXBHUEWBX-UHFFFAOYSA-N
Synonym ethyl 2-bromonicotinate, 2-bromonicotinic acid ethyl ester, 2-bromonicotinic ethyl ester, ethyl 2-bromonicotite, pubchem12886, ethyl 2-bromanylpyridine-3-carboxylate, 2-bromo-nicotinic acid ethyl ester, ethyl 2-bromo-3-pyridinecarboxylate, 2-bromo-3-pyridinecarboxylic acid ethyl ester, 2-bromo-pyridine-3-carboxylic acid ethyl ester
PubChem CID 1415877
IUPAC Name ethyl 2-bromopyridine-3-carboxylate
SMILES CCOC(=O)C1=C(N=CC=C1)Br

Specifications

Quantity 250 mg
Formula Weight 230.07
Percent Purity 96%
Chemical Name or Material Ethyl 2-bromonicotinate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.