missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-(bromomethyl)acrylate, 97%

Product Code. 10398700 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10398700 5 g Glass bottle 5g
10087292 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10398700 Supplier Thermo Scientific Acros Supplier No. 235360050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 17435-72-2 | C6H9BrO2 | 193.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17435-72-2
Molecular Formula C6H9BrO2
Molecular Weight (g/mol) 193.04
MDL Number MFCD00031518
InChI Key MTCMFVTVXAOHNQ-UHFFFAOYSA-N
Synonym ethyl 2-bromomethyl acrylate, ethyl 2-bromomethyl prop-2-enoate, 2-bromomethyl acrylic acid ethyl ester, ethyl alpha-bromomethyl acrylate, 2-propenoic acid, 2-bromomethyl-, ethyl ester, 2-bromomethyl-acrylic acid ethyl ester, ethyl-2-bromomethylacrylate, ethyl 2-bromomethylacrylate, acmc-1bo78, ksc181c2b
PubChem CID 310620
IUPAC Name ethyl 2-(bromomethyl)prop-2-enoate
SMILES CCOC(=O)C(=C)CBr

Specifications

Density 1.3800g/mL
Boiling Point 86°C (20.0 mmHg)
Flash Point 81°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH2=C(CH2Br)COOC2H5
Refractive Index 1.4770 to 1.479
Quantity 5 g
Fieser 13,322; 14,162
Specific Gravity 1.38
Formula Weight 193.05
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Ethyl 2-(bromomethyl)acrylate, 97%
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl 2-(bromomethyl)acrylate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.