missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-bromoisovalerate, 97%

Product Code. 11430727
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11430727 5 g 5g
11440727 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11430727 Supplier Thermo Scientific Alfa Aesar Supplier No. B22525.06
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 609-12-1 | C7H13BrO2 | 209.083 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 609-12-1
Molecular Formula C7H13BrO2
Molecular Weight (g/mol) 209.083
MDL Number MFCD00026855
InChI Key WNFUWONOILPKNX-UHFFFAOYSA-N
Synonym ethyl 2-bromoisovalerate, ethyl 2-bromo-3-methylbutyrate, butanoic acid, 2-bromo-3-methyl-, ethyl ester, ethyl .alpha.-bromoisovalerate, 2-bromoisovaleric acid ethyl ester, butyric acid, 2-bromo-3-methyl-, ethyl ester, ethyl 2-bromo-3-methyl-butanoate, .alpha.-bromoisovaleric acid ethyl ester, ethyl, a-bromoisovalerate, ethyl 2-bromo isovalerate
PubChem CID 79068
IUPAC Name ethyl 2-bromo-3-methylbutanoate
SMILES CCOC(=O)C(C(C)C)Br

Specifications

Boiling Point 185°C to 186°C
Flash Point 65°C (149°F)
Odor Pungent
Quantity 5 g
UN Number UN3265
Beilstein 1099039
Formula Weight 209.09
Percent Purity 97%
Chemical Name or Material Ethyl 2-bromoisovalerate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.