missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-bromoisobutyrate, 98%

Product Code. 10142910 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
500 g
Packaging:
Glass bottle
Unit Size:
500g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10142910 500 g Glass bottle 500g
10560581 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10142910 Supplier Thermo Scientific Acros Supplier No. 118135000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 600-00-0 | C6H11BrO2 | 195.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 600-00-0
Molecular Formula C6H11BrO2
Molecular Weight (g/mol) 195.06
MDL Number MFCD00000123
InChI Key IOLQWGVDEFWYNP-UHFFFAOYSA-N
Synonym ethyl 2-bromoisobutyrate, ethyl 2-bromo-2-methylpropionate, ethyl alpha-bromoisobutyrate, ethyl .alpha.-bromoisobutyrate, propanoic acid, 2-bromo-2-methyl-, ethyl ester, 2-bromo-2-methylpropanoic acid, ethyl ester, ethyl-2-bromoisobutyrate, propanoic acid, bromo-2-methyl-, ethyl ester, ethyl 2-bromo-2-methyl-propanoate, propionic acid, 2-bromo-2-methyl-, ethyl ester
PubChem CID 11745
IUPAC Name ethyl 2-bromo-2-methylpropanoate
SMILES CCOC(=O)C(C)(C)Br

Specifications

Color Colorless to Yellow
Density 1.3290g/mL
Boiling Point 161°C to 163°C
Flash Point 60°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2C(Br)CO2C2H5
Refractive Index 1.4430 to 1.446
Quantity 500 g
Beilstein 02, 296
Specific Gravity 1.329
Formula Weight 195.06
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethyl 2-bromoisobutyrate, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.