missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-bromo-4-methylthiazole-5-carboxylate, 97%, Thermo Scientific™

Product Code. 15498297
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15498297 1 g 1g
15408307 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15498297 Supplier Thermo Scientific Alfa Aesar Supplier No. H51833.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It finds its application in the preparation of thiazole-4- and -5-carboxylates, and an infrared study of their rotational isomers and in the discovery of thiazolylpyridinone SCD1 inhibitors with preferential liver distribution and reduced mechanism-based adverse effects.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22900-83-0
Molecular Formula C7H8BrNO2S
Molecular Weight (g/mol) 250.11
MDL Number MFCD03791227
InChI Key CFBIOWPDDZPIDP-UHFFFAOYSA-N
Synonym ethyl 2-bromo-4-methylthiazole-5-carboxylate, 5-thiazolecarboxylic acid, 2-bromo-4-methyl-, ethyl ester, 2-bromo-4-methyl-thiazole-5-carboxylic acid ethyl ester, ethyl 2-bromo-4-methyl-5-thiazolecarboxylate, 2-bromo-4-methyl-5-thiazolecarboxylic acid ethyl ester, 2-bromo-4-methylthiazole-5-carboxylic acid ethyl ester, maybridge3_004543, ksc207i5t, ethyl2-bromo-4-methyl-1,3-thiazole-5-carboxylate, ethylbromomethylthiazolecarboxylate
PubChem CID 2824057
IUPAC Name ethyl 2-bromo-4-methyl-1,3-thiazole-5-carboxylate
SMILES CCOC(=O)C1=C(N=C(S1)Br)C

Specifications

Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 250.12
Percent Purity 97%
Chemical Name or Material Ethyl 2-bromo-4-methylthiazole-5-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.