missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-bromo-3-methylbutyrate, 95%

Product Code. 11934941 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass Bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11934941 50 g Glass Bottle 50g
10065780 10 g Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11934941 Supplier Thermo Scientific Acros Supplier No. 409790500
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 609-12-1 | C7H13BrO2 | 209.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 609-12-1
Molecular Formula C7H13BrO2
Molecular Weight (g/mol) 209.08
InChI Key WNFUWONOILPKNX-UHFFFAOYSA-N
Synonym ethyl 2-bromoisovalerate, ethyl 2-bromo-3-methylbutyrate, butanoic acid, 2-bromo-3-methyl-, ethyl ester, ethyl .alpha.-bromoisovalerate, 2-bromoisovaleric acid ethyl ester, butyric acid, 2-bromo-3-methyl-, ethyl ester, ethyl 2-bromo-3-methyl-butanoate, .alpha.-bromoisovaleric acid ethyl ester, ethyl, a-bromoisovalerate, ethyl 2-bromo isovalerate
PubChem CID 79068
IUPAC Name ethyl 2-bromo-3-methylbutanoate
SMILES CCOC(=O)C(C(C)C)Br

Specifications

Color Gray
Boiling Point 77.0°C (12.0 mmHg)
Flash Point 65°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Linear Formula (CH3)2CHCHBrCOOC2H5
Refractive Index 1.4485 to 1.4505
Quantity 50 g
Formula Weight 209.08
Percent Purity 95%
Physical Form Crystals, Powder or Flakes
Chemical Name or Material Ethyl 2-bromo-3-methylbutyrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.