missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (2-Azidoethoxy)acetoacetate, TRC

Product Code. 30382155
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30382155 250 mg 250mg
30388427 500 mg 500mg
30378496 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30382155 Supplier Toronto Research Chemicals Supplier No. TRCE899555250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88150-45-2
Molecular Formula C8 H13 N3 O4
Molecular Weight (g/mol) 215.21
Synonym Butanoic acid, 4-(2-azidoethoxy)-3-oxo-, ethyl ester, 4-(2-Azidoethoxy)-3-oxobutanoic acid ethyl ester
IUPAC Name ethyl 4-(2-azidoethoxy)-3-oxobutanoate
SMILES CCOC(=O)CC(=O)COCCN=[N+]=[N-]

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C8H13N3O4/c1-2-15-8(13)5-7(12)6-14-4-3-10-11-9/h2-6H2,1H3
Formula Weight 215.0906
Chemical Name or Material Ethyl (2-Azidoethoxy)acetoacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.