missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-aminopyridine-4-carboxylate, 97%

Product Code. 15487788
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15487788 5 g 5g
15497788 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15487788 Supplier Thermo Scientific Alfa Aesar Supplier No. H61280.06
 Restricted Item
Estimated Shipment: 15-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 13362-30-6 | C8H10N2O2 | 166.18 g/mol

Ethyl 2-aminopyridine-4-carboxylate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 2-aminopyridine-4-carboxylate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13362-30-6
Molecular Formula C8H10N2O2
Molecular Weight (g/mol) 166.18
MDL Number MFCD03791260
InChI Key XVBZFXZNJAFCHL-UHFFFAOYSA-N
Synonym ethyl 2-aminoisonicotinate, 2-amino-isonicotinic acid ethyl ester, ethyl 2-amino-4-pyridinecarboxylate, 2-aminoisonicotinic acid ethyl ester, 2-amino-4-ethoxycarbonyl pyridine, 4-pyridinecarboxylic acid, 2-amino-, ethyl ester, 2-amino-4-pyridinecarboxylic acid ethyl ester, 2-aminopyridine-4-carboxylic acid ethyl ester, pubchem16599, acmc-209bt6
PubChem CID 7015521
IUPAC Name ethyl 2-aminopyridine-4-carboxylate
SMILES CCOC(=O)C1=CC(=NC=C1)N

Specifications

Color White
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 166.18
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Ethyl 2-aminopyridine-4-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.