missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-aminonicotinate, 98%

Product Code. 15432389
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15432389 5 g 5g
15442389 25 g 25g
15452389 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15432389 Supplier Thermo Scientific Alfa Aesar Supplier No. H64957.06
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 13362-26-0 | C8H10N2O2 | 166.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13362-26-0
Molecular Formula C8H10N2O2
Molecular Weight (g/mol) 166.18
MDL Number MFCD01569452
InChI Key KIAGEDYOPMHRRB-UHFFFAOYSA-N
PubChem CID 12433260
IUPAC Name ethyl 2-aminopyridine-3-carboxylate
SMILES CCOC(=O)C1=C(N=CC=C1)N

Specifications

Melting Point 90°C to 96°C (literature)
Quantity 5 g
Beilstein 124680
Synonym ethyl 2-aminonicotinate, 2-amino-nicotinic acid ethyl ester, ethyl2-aminonicotinate, 2-aminonicotinic acid ethyl ester, 3-pyridinecarboxylic acid, 2-amino-, ethyl ester, 2-amino nicotinic acid ethyl ester, 2-aminopyridine-3-carboxylic acid ethyl ester, pubchem12921, rarechem al bi 1234, buttpark 23\07-65
Formula Weight 166.18
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Ethyl 2-aminonicotinate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.