missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-aminobenzothiazole-6-carboxylate, 97%, Thermo Scientific Chemicals

Product Code. 15472429
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50850-93-6
Molecular Formula C10H10N2O2S
Molecular Weight (g/mol) 222.262
MDL Number MFCD00102724
InChI Key VYJSGJXWKSDUSG-UHFFFAOYSA-N
PubChem CID 601008
IUPAC Name ethyl 2-amino-1,3-benzothiazole-6-carboxylate
SMILES CCOC(=O)C1=CC2=C(C=C1)N=C(S2)N

Specifications

Melting Point 241°C to 245°C
Quantity 25 g
Beilstein 185097
Synonym ethyl 2-aminobenzo d thiazole-6-carboxylate, ethyl 2-amino-benzothiazole-6-carboxylate, ethyl 2-aminobenzothiazole-6-carboxylate, 2-amino-benzothiazole-6-carboxylic acid ethyl ester, 6-benzothiazolecarboxylic acid, 2-amino-, ethyl ester, 2-amino-6-ethoxycarbonylbenzothiazole, ethyl2-amino-benzothiazole-6-carboxylate, 2-aminobenzothiazole-6-carboxylic acid ethyl ester, 2-amino benzo thiazole-6-carboxylic acid ethyl ester, 3kid
Formula Weight 222.26
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Ethyl 2-aminobenzothiazole-6-carboxylate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'