missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-amino-6-Boc-4,7-dihydrothieno[2,3-c]pyridine-3(5H)-carboxylate, 97%, Thermo Scientific™

Product Code. 15405205
Change view
Click to view available options
Quantity:
250 mg
5 g
Unit Size:
250mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15405205 250 mg 250mg
15438865 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15405205 Supplier Thermo Scientific Alfa Aesar Supplier No. H34279.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 193537-14-3
Molecular Formula C15H22N2O4S
Molecular Weight (g/mol) 326.411
MDL Number MFCD05664039
InChI Key XSXVOVXVHBSSSN-UHFFFAOYSA-N
Synonym ethyl 6-boc-2-amino-4,7-dihydro-5h-thieno 2,3-c pyridine-3-carboxylate, 2-amino-4,7-dihydro-5h-thieno 2,3-c pyridine-3,6-dicarboxylic acid 6-tert butyl ester 3-ethyl ester, 6-tert-butyl 3-ethyl 2-amino-4,5-dihydrothieno 2,3-c pyridine-3,6 7h-dicarboxylate, 6-tert-butyl 3-ethyl 2-amino-4h,5h,6h,7h-thieno 2,3-c pyridine-3,6-dicarboxylate, 6-tert-butyl 3-ethyl 2-amino-4,7-dihydrothieno 2,3-c pyridine-3,6 5h-dicarboxylate, 2-amino-6-boc-4,5,6,7-tetrahydro-thieno 2,3-c pyridine-3-carboxylic acid ethyl ester, 2-amino-4,7-dihydro-5h-thieno 2,3-c pyridine-3,6-dicarboxylic acid 6-tert-butyl ester 3-ethyl ester, 6-tert-butyl 3-ethyl 2-amino-4h,5h,7h-thieno 2,3-c pyridine-3,6-dicarboxylate, ethyl 2-amino-4,5,6,7-tetrahydrothieno 2,3-c pyridine-3-carboxylate, n-boc protected
PubChem CID 2794740
IUPAC Name 6-O-tert-butyl 3-O-ethyl 2-amino-5,7-dihydro-4H-thieno[2,3-c]pyridine-3,6-dicarboxylate
SMILES CCOC(=O)C1=C(SC2=C1CCN(C2)C(=O)OC(C)(C)C)N

Specifications

Melting Point 151°C to 153°C
Quantity 250 mg
Formula Weight 326.42
Percent Purity 97%
Chemical Name or Material Ethyl 2-amino-6-Boc-4,7-dihydrothieno[2,3-c]pyridine-3(5H)-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.