missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-amino-5-phenylthiophene-3-carboxylate, 97%

Product Code. 15496687
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15496687 1 g 1g
15406697 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15496687 Supplier Thermo Scientific Alfa Aesar Supplier No. H33823.03
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4815-34-3 | C13H13NO2S | 247.312 g/mol

Ethyl 2-amino-5-phenylthiophene-3-carboxylate is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 2-amino-5-phenylthiophene-3-carboxylate is used as a pharmaceutical intermediate.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4815-34-3
Molecular Formula C13H13NO2S
Molecular Weight (g/mol) 247.312
MDL Number MFCD01829801
InChI Key WIVNPGXPJBBZQH-UHFFFAOYSA-N
Synonym ethyl 2-amino-5-phenyl-3-thiophenecarboxylate, 2-amino-5-phenyl-thiophene-3-carboxylic acid ethyl ester, 2-amino-5-phenylthiophene-3-carboxylic acid ethyl ester, ethyl2-amino-5-phenylthiophene-3-carboxylate, 2-amino-5-phenyl-3-thiophenecarboxylic acid ethyl ester, enamine_004942, 2-amino-3-ethoxycarbonyl-5-phenylthiophene, ethyl 2-azanyl-5-phenyl-thiophene-3-carboxylate, 2-amino-5-phenyl thiophene-3-carboxylic ethyl ester, 2-amino-5-phenyl-3-thiophenecarboxylic acid, ethyl ester
PubChem CID 638860
IUPAC Name ethyl 2-amino-5-phenylthiophene-3-carboxylate
SMILES CCOC(=O)C1=C(SC(=C1)C2=CC=CC=C2)N

Specifications

Melting Point 121°C to 123°C
Quantity 1 g
Formula Weight 247.32
Percent Purity 97%
Chemical Name or Material Ethyl 2-amino-5-phenylthiophene-3-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.