missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carboxylate, 96%

Product Code. 15498355
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15498355 25 g 25g
15488355 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15498355 Supplier Thermo Scientific Alfa Aesar Supplier No. H33196.14
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 4815-29-6 | C10H13NO2S | 211.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4815-29-6
Molecular Formula C10H13NO2S
Molecular Weight (g/mol) 211.28
MDL Number MFCD00102063
InChI Key BOJXCJDYZJSPMZ-UHFFFAOYSA-N
Synonym ethyl 2-amino-5,6-dihydro-4h-cyclopenta b thiophene-3-carboxylate, 2-amino-5,6-dihydro-4h-cyclopenta b thiophene-3-carboxylic acid ethyl ester, ethyl 2-amino-4h,5h,6h-cyclopenta b thiophene-3-carboxylate, ethyl 2-aminocyclopenta b thiophene-3-carboxylate, ethyl 2-amino-5,6-dihydro-4h-cyclopenta b-thiophene-3-carboxylate, 4h-cyclopenta b thiophene-3-carboxylic acid, 2-amino-5,6-dihydro-, ethyl ester, ethyl 2-amino-4,5,6-trihydrocyclopenta 1,2-b thiophene-3-carboxylate, enamine_004067, ethyl 2-aminocyclopentathiophene-3-carboxylate
PubChem CID 264105
IUPAC Name ethyl 2-amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carboxylate
SMILES CCOC(=O)C1=C(N)SC2=C1CCC2

Specifications

Melting Point 95°C to 96°C
Quantity 25 g
Formula Weight 211.28
Percent Purity 96%
Chemical Name or Material Ethyl 2-amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.