missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-amino-4-phenyl-5-thiazolecarboxylate, 99%, Thermo Scientific™

Product Code. 10072000 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 g
Packaging:
Glass bottle
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10072000 2.5 g Glass bottle 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10072000 Supplier Thermo Scientific Acros Supplier No. 162220025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 64399-23-1
Melting Point 169°C to 172°C
Color Brown
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C12H12N2O2S
MDL Number MFCD00022452
Quantity 2.5 g
Synonym ethyl 2-amino-4-phenylthiazole-5-carboxylate, ethyl 2-amino-4-phenyl-5-thiazolecarboxylate, 5-thiazolecarboxylic acid, 2-amino-4-phenyl-, ethyl ester, 2-amino-4-phenylthiazole-5-carboxylic acid ethyl ester, acmc-209nlv, 2-amino-4-phenyl-5-carboethoxythiazole, ethyl 2-amino-4-phenylthiazol-5-carboxylate, ethyl2-amino-4-phenylthiazole-5-carboxylate, ethyl 2-amino-4-phenyl-thiazole-5-carboxylate, ethyl 2-amino-4-phenylthiazol-5-yl-carboxylate
InChI Key OZMXFXOHCUEEPD-UHFFFAOYSA-N
SMILES CCOC(=O)C1=C(N=C(S1)N)C2=CC=CC=C2
IUPAC Name ethyl 2-amino-4-phenyl-1,3-thiazole-5-carboxylate
Molecular Weight (g/mol) 248.3
PubChem CID 73239
Formula Weight 248.3
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 2-amino-4-phenyl-5-thiazolecarboxylate, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.