missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-amino-4-methylthiophene-3-carboxylate, 98+%

Product Code. 11419712
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11419712 5 g 5g
11409712 1 g 1g
11429712 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11419712 Supplier Thermo Scientific Alfa Aesar Supplier No. A10790.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 43088-42-2 | C8H11NO2S | 185.241 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 43088-42-2
Molecular Formula C8H11NO2S
Molecular Weight (g/mol) 185.241
MDL Number MFCD00051669
InChI Key ILYCZKOBLRJJSW-UHFFFAOYSA-N
Synonym 2-amino-4-methylthiophene-3-carboxylic acid ethyl ester, 2-amino-4-methyl-thiophene-3-carboxylic acid ethyl ester, 3-thiophenecarboxylic acid, 2-amino-4-methyl-, ethyl ester, ethyl2-amino-4-methylthiophene-3-carboxylate, ethyl 2-amino-4-methyl-3-thiophenecarboxylate, acmc-20a9ux, maybridge1_003755, 2-amino-4-methyl-3-ethoxycarbonyl-thiophene, ethyl 2-amino4-methylthiophene-3-carboxylate, 2-amino-3-ethoxycarbonyl-4-methylthiophene
PubChem CID 521132
IUPAC Name ethyl 2-amino-4-methylthiophene-3-carboxylate
SMILES CCOC(=O)C1=C(SC=C1C)N

Specifications

Melting Point 79°C to 83°C
Quantity 5 g
Beilstein 1366073
Formula Weight 185.25
Percent Purity ≥98%
Chemical Name or Material Ethyl 2-amino-4-methylthiophene-3-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.