missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-amino-4-methylthiazole-5-carboxylate, 97%

Product Code. p-7023391
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11432928

Brand: Thermo Scientific Alfa Aesar A16896.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7210-76-6 | C7H10N2O2S | 186.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7210-76-6
Molecular Formula C7H10N2O2S
Molecular Weight (g/mol) 186.23
MDL Number MFCD00123414
InChI Key WZHUPCREDVWLKC-UHFFFAOYSA-N
PubChem CID 343747
IUPAC Name ethyl 2-amino-4-methyl-1,3-thiazole-5-carboxylate
SMILES CCOC(=O)C1=C(C)N=C(N)S1

Specifications

Melting Point 172°C to 176°C
Odor Odorless
Quantity 5 g
Synonym ethyl 2-amino-4-methylthiazole-5-carboxylate, ethyl2-amino-4-methylthiazole-5-carboxylate, 2-amino-4-methyl-thiazole-5-carboxylic acid ethyl ester, 5-thiazolecarboxylic acid, 2-amino-4-methyl-, ethyl ester, ethyl 2-amino-4-methyl thiazole-5-carboxylate, ethyl 2-amino-4-methyl-thiazole-5-carboxylate, 2-amino-4-methyl-5-thiazolecarboxylic acid ethyl ester, 2-amino-4-methyl-5-ethoxycarbonyl thiazole, 2-amino-4-methyl-5-ethoxycarbonyl-thiazole, ethyl 2-amino-4-methyl-5-thiazolecarboxylate
Formula Weight 186.22
Percent Purity 97%
Chemical Name or Material Ethyl 2-amino-4-methylthiazole-5-carboxylate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.