missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-amino-4-methylthiazole-5-carboxylate, 97%

Product Code. 11442928
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11442928 25 g 25g
11432928 5 g 5g
11458716 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11442928 Supplier Thermo Scientific Alfa Aesar Supplier No. A16896.14
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 7210-76-6 | C7H10N2O2S | 186.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7210-76-6
Molecular Formula C7H10N2O2S
Molecular Weight (g/mol) 186.23
MDL Number MFCD00123414
InChI Key WZHUPCREDVWLKC-UHFFFAOYSA-N
PubChem CID 343747
IUPAC Name ethyl 2-amino-4-methyl-1,3-thiazole-5-carboxylate
SMILES CCOC(=O)C1=C(C)N=C(N)S1

Specifications

Melting Point 172°C to 176°C
Odor Odorless
Quantity 25 g
Synonym ethyl 2-amino-4-methylthiazole-5-carboxylate, ethyl2-amino-4-methylthiazole-5-carboxylate, 2-amino-4-methyl-thiazole-5-carboxylic acid ethyl ester, 5-thiazolecarboxylic acid, 2-amino-4-methyl-, ethyl ester, ethyl 2-amino-4-methyl thiazole-5-carboxylate, ethyl 2-amino-4-methyl-thiazole-5-carboxylate, 2-amino-4-methyl-5-thiazolecarboxylic acid ethyl ester, 2-amino-4-methyl-5-ethoxycarbonyl thiazole, 2-amino-4-methyl-5-ethoxycarbonyl-thiazole, ethyl 2-amino-4-methyl-5-thiazolecarboxylate
Formula Weight 186.22
Percent Purity 97%
Chemical Name or Material Ethyl 2-amino-4-methylthiazole-5-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.