missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-amino-4-hydroxypyrimidine-5-carboxylate, 95%

Product Code. p-7044473
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11306946

Brand: Thermo Scientific Alfa Aesar L06654.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15400-53-0 | C7H9N3O3 | 183.167 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15400-53-0
Molecular Formula C7H9N3O3
Molecular Weight (g/mol) 183.167
MDL Number MFCD00039708
InChI Key HRRHGLKNOJHIGY-UHFFFAOYSA-N
Synonym ethyl 2-amino-4-hydroxypyrimidine-5-carboxylate, 2-amino-5-carboethoxy-4-hydroxypyrimidine, 2-amino-4-hydroxypyrimidine-5-carboxylic acid ethyl ester, ethyl 2-amino-1,4-dihydro-4-oxopyrimidine-5-carboxylate, 2-amino-4-hydroxy-pyrimidine-5-carboxylic acid ethyl ester, 2-amino-6-oxo-1,6-dihydro-pyrimidine-5-carboxylic acid ethyl ester, 5-pyrimidinecarboxylic acid, 2-amino-1,4-dihydro-4-oxo-, ethyl ester, ethyl 2-amino-4-oxo-3h-pyrimidine-5-carboxylate, 1rry, acmc-209xd5
PubChem CID 84894
ChEBI CHEBI:39721
IUPAC Name ethyl 2-amino-6-oxo-1H-pyrimidine-5-carboxylate
SMILES CCOC(=O)C1=CN=C(NC1=O)N

Specifications

Melting Point ∽300°C (decomposition)
Odor Odorless
Quantity 1 g
Beilstein 165835
Formula Weight 183.17
Percent Purity 95%
Chemical Name or Material Ethyl 2-amino-4-hydroxypyrimidine-5-carboxylate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.