missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-amino-4,5-dimethyl-3-thiophenecarboxylate, 97%, Thermo Scientific™

Product Code. 15296008 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15296008 1 g 1g
15206018 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15296008 Supplier Thermo Scientific Acros Supplier No. 450300010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 4815-24-1
Molecular Formula C9H13NO2S
Molecular Weight (g/mol) 199.27
InChI Key JYSDXODDWAQWJR-UHFFFAOYSA-N
Synonym ethyl 2-amino-4,5-dimethyl3-thenoate, ethyl 2-amino-4,5-dimethyl-3-thiophenecarboxylate, 2-amino-4,5-dimethylthiophene-3-carboxylic acid ethyl ester, 2-amino-4,5-dimethyl-thiophene-3-carboxylic acid ethyl ester, 3-thiophenecarboxylic acid, 2-amino-4,5-dimethyl-, ethyl ester, 2-amino-3-carbethoxy-4,5-dimethylthiophene, 2-amino-4,5-dimethyl-3-ethoxycarbonyl-thiophene, ethyl-2-amino-4,5-dimethylthiophene-3-carboxylate, ethyl2-amino-4,5-dimethylthiophene-3-carboxylate, ethyl 2-amino-4,5-dimethyl-thiophene-3-carboxylate
PubChem CID 78536
IUPAC Name ethyl 2-amino-4,5-dimethylthiophene-3-carboxylate
SMILES CCOC(=O)C1=C(SC(=C1C)C)N

Specifications

Melting Point 90.0°C to 91.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 199.27
Percent Purity 97%
Chemical Name or Material Ethyl 2-amino-4,5-dimethyl-3-thiophenecarboxylate
compliance-icons
Product Identifier
  • Ethyl 2-amino-4,5-dimethyl-3-thiophenecarboxylate, 97%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.