missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-amino-4-(4-chlorophenyl)thiophene-3-carboxylate, 96%, Thermo Scientific™

Product Code. 15488925
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15488925 1 g 1g
15498925 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15488925 Supplier Thermo Scientific Alfa Aesar Supplier No. H34427.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used in the design, synthesis and biological evaluation of novel condensed fuoropyrimidines as anti-hyperlipidemic agents. Synthesis of 2-chloromethyl-5-(4- chlorophenyl) thieno [2,3-d]pyrimidin- 4(3H)-one. A stream of dry hydrogen chloride gas was bubbled through an ice-cold mixture of ethyl 2-amino-4-(4-chlorophenyl)thiophene 3- carboxylate.

Notes
Incompatible with strong oxidizing agents. Store at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65234-09-5
Molecular Formula C13H12ClNO2S
Molecular Weight (g/mol) 281.75
MDL Number MFCD01038372
InChI Key YHJWJJKFFNCDCK-UHFFFAOYSA-N
Synonym ethyl 2-amino-4-4-chlorophenyl thiophene-3-carboxylate, 2-amino-4-4-chloro-phenyl-thiophene-3-carboxylicacidethylester, 2-amino-4-4-chlorophenyl thiophene-3-carboxylic acid ethyl ester, 2-amino-4-4-chloro-phenyl-thiophene-3-carboxylic acid ethyl ester, us8889672, apogee a, 2-amino-4-4-chlorophenyl thiophene-3-carboxylate, ethyl 2-amino-4-4-chlorophenyl-3-thiophenecarboxylate, ethyl 2-amino-4-4-chlorophenyl-3-thiophenecarboxylate #, ethyl 2-amino-4-4-chlorophenyl-thiophene-3-carboxylate, 3-thiophenecarboxylicacid,2-amino-4-4-chlorophenyl-,ethyl ester
PubChem CID 616598
IUPAC Name ethyl 2-amino-4-(4-chlorophenyl)thiophene-3-carboxylate
SMILES CCOC(=O)C1=C(N)SC=C1C1=CC=C(Cl)C=C1

Specifications

Melting Point 110°C to 114°C
Quantity 1 g
Formula Weight 281.76
Percent Purity 96%
Chemical Name or Material Ethyl 2-amino-4-(4-chlorophenyl)thiophene-3-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.