missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-amidinoacetate hydrochloride, 95%

Product Code. 15411419
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15411419 5 g 5g
15401419 1 g 1g
15421419 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15411419 Supplier Thermo Scientific Alfa Aesar Supplier No. H64288.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 57508-48-2 | C5H11ClN2O2 | 166.605 g/mol

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57508-48-2
Molecular Formula C5H11ClN2O2
Molecular Weight (g/mol) 166.605
MDL Number MFCD06797615
InChI Key VOHFLYOSVGWQOS-UHFFFAOYSA-N
Synonym ethyl 3-amino-3-iminopropanoate hydrochloride, ethyl 2-amidinoacetate hydrochloride, 3-amino-3-iminopropanoic acid ethyl ester hydrochloride, ethyl 2-carbamimidoylacetate hydrochloride, 3-amino-3-imino-propanoic acid, ethyl ester hcl, carbamimidoyl-acetic acid ethyl ester hydrochloride, ethyl 2-amidinoacetate hcl, ethyl 3-amino-3-iminopropanate hydrochloride, ethyl 3-amino-3-iminopropanoate hcl, 3-amino-3-iminopropionic acid ethyl ester hydrochloride
PubChem CID 15555354
IUPAC Name ethyl 3-amino-3-iminopropanoate;hydrochloride
SMILES CCOC(=O)CC(=N)N.Cl

Specifications

Melting Point 105°C to 108°C
Quantity 5 g
Beilstein 3694030
Formula Weight 166.6
Percent Purity 95%
Physical Form Solid
Chemical Name or Material Ethyl 2-amidinoacetate hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.