missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-Acetylhexanoate, TRC

Product Code. 30384552
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30384552 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30384552 Supplier Toronto Research Chemicals Supplier No. TRCE89642350MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl 2-Acetylhexanoate
CAS 1540-29-0
Molecular Formula C10H18O3
Quantity 50 mg
Synonym Ethyl 2-Butyl-3-oxobutanoate, Ethyl 2-Butylacetoacetate, Ethyl 2-Butylacetylacetate, Ethyl alpha-Butylacetoacetate, NSC 11771, NSC 25322, NSC 404202, 2-Acetylhexanoic Acid Ethyl Ester, 3-Carbethoxy-2-heptanone
InChI Formula InChI=1S/C10H18O3/c1-4-6-7-9(8(3)11)10(12)13-5-2/h9H,4-7H2,1-3H3
SMILES CCCCC(C(=O)C)C(=O)OCC
IUPAC Name ethyl 2-acetylhexanoate
Molecular Weight (g/mol) 186.25
Formula Weight 186.1256
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.