missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-Acetamido-4-(4-octylphenyl)butanoate, TRC

Product Code. 30386647
Change view
Click to view available options
Quantity:
0.5 mg
5 mg
25 mg
Unit Size:
0.5mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30386647 5 mg 5mg
30369037 0.5 mg 0.5mg
30386411 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30386647 Supplier Toronto Research Chemicals Supplier No. TRCE9380435MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl 2-Acetamido-4-(4-octylphenyl)butanoate
Analyte or Component Names Ethyl 2-Acetamido-4-(4-octylphenyl)butanoate
Shipping Condition Room Temperature
Molecular Formula C22 H35 N O3
InChI Formula InChI=1S/C22H35NO3/c1-4-6-7-8-9-10-11-19-12-14-20(15-13-19)16-17-21(23-18(3)24)22(25)26-5-2/h12-15,21H,4-11,16-17H2,1-3H3,(H,23,24)
Solution Type Neat
SMILES CCCCCCCCc1ccc(CCC(NC(=O)C)C(=O)OCC)cc1
IUPAC Name ethyl 2-acetamido-4-(4-octylphenyl)butanoate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 361.518
Quantity 5 mg
Formula Weight 361.262
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.