missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-(6-Amino-2,3-dichlorobenzyl)glycine-13C2(Anagrelide Impurity A), TRC

Product Code. 30380029
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380029 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30380029 Supplier Toronto Research Chemicals Supplier No. TRCE8981821MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl 2-(6-Amino-2,3-dichlorobenzyl)glycine-13C2(Anagrelide Impurity A)
CAS 1286968-94-2
Molecular Formula C913C2H14Cl2N2O2
Quantity 1 mg
Synonym N-[(6-Amino-2,3-dichlorophenyl)methyl]glycine Ethyl Ester-13C2, Ethyl (2-Amino-5,6-dichlorobenzyl)aminoacetate-13C2, Ethyl N-(6-Amino-2,3-dichlorobenzyl)glycine-13C2, USP Anagrelide Related Impurity A-13C2
InChI Formula InChI=1S/C11H14Cl2N2O2/c1-2-17-10(16)6-15-5-7-9(14)4-3-8(12)11(7)13/h3-4,15H,2,5-6,14H2,1H3/i6+1,10+1
SMILES CCO[13C](=O)[13CH2]NCc1c(N)ccc(Cl)c1Cl
Molecular Weight (g/mol) 279.13
Formula Weight 278.0499
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.