missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2,4-dioxovalerate, 98%, Thermo Scientific Chemicals

Product Code. 10549633 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10549633 25 g Glass bottle 25g
10082640 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10549633 Supplier Thermo Scientific Acros Supplier No. 214010250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 615-79-2
Molecular Formula C7H10O4
Molecular Weight (g/mol) 158.15
MDL Number MFCD00009124
InChI Key OYQVQWIASIXXRT-UHFFFAOYSA-N
Synonym ethyl 2,4-dioxovalerate, ethyl acetopyruvate, pentanoic acid, 2,4-dioxo-, ethyl ester, ethyl2,4-dioxovalerate, 2,4-dioxo-pentanoic acid ethyl ester, 2,4-dioxopentanoic acid ethyl ester, 2,4-dioxo-pentanoic-acid-ethyl-ester, 2,4-dioxo-pentanoicacid ethylester, ethyl 2,4-dioxo-valerate, ethyl 2,4-dioxo-pentanoate
PubChem CID 69208
IUPAC Name ethyl 2,4-dioxopentanoate
SMILES CCOC(=O)C(=O)CC(C)=O

Specifications

Melting Point 16.0°C to 18.0°C
Color Yellow
Density 1.1200g/mL
Boiling Point 101.0°C to 103.0°C (12.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3COCH2COCO2C2H5
Refractive Index 1.4730 to 1.4750
Quantity 25 g
Beilstein 03, 747
Specific Gravity 1.12
Formula Weight 158.15
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethyl 2,4-dioxovalerate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.