missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 2,4-dichloropyrimidine-5-carboxylate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
It's employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It′s employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
It′s employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications
Specifications
Melting Point | 36°C to 37°C |
Color | White |
Quantity | 5 g |
Beilstein | 152173 |
Synonym | ethyl 2,4-dichloro-5-pyrimidinecarboxylate, 5-pyrimidinecarboxylic acid, 2,4-dichloro-, ethyl ester, ethyl 2,4-dichloro-pyrimidine-5-carboxylate, 2,4-dichloro-pyrimidine-5-carboxylic acid ethyl ester, 2,4-dichloro-5-ethoxycarbonylpyrimidine, 2,4,-dichloro-5-pyrimidinecarboxylic acid ethyl ester, 2,4-dichloro-5-pyrimidinecarboxylic acid ethyl ester, 2,4-dichloropyrimidine-5-carboxylic acid ethyl ester, pubchem24010, ethyl2,4-dichloro-5-pyrimidinecarboxylate |
Solubility Information | Slightly soluble in water. |
Formula Weight | 221.04 |
Percent Purity | 98% |
Physical Form | Powder |
Chemical Name or Material | Ethyl 2,4-dichloropyrimidine-5-carboxylate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'